Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2,5-Bis(1-naphthyl)-1,3,4-oxadiazole ≥98.0% (by HPLC, total nitrogen)

Supplier: TCI America


D2757-5G
TCD2757-5GEA 139.41 USD InStock
TCD2757-5G
2,5-Bis(1-naphthyl)-1,3,4-oxadiazole ≥98.0% (by HPLC, total nitrogen)
2,5-Bis(1-naphthyl)-1,3,4-oxadiazole
CAS Number: 905-62-4
MDL Number: MFCD00045582
Molecular Formula: C22H14N2O
Molecular Weight: 322.37
Purity/Analysis Method: 98.0% (N,HPLC)
Form: Crystal
Melting point (°C): 179


Formula: C₂₂H₁₄N₂O
MW: 322,37
Melting Pt: 179 °C
MDL Number: MFCD00045582
CAS Number: 905-62-4
EINECS: 212-995-0
Order Now
Specification Test Results
Purity/Analysis Method 98.0% (N,HPLC)


 

ORDER

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty