Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2,3-Dibromo-1,1,3,3-tetrafluoropropene ≥95.0%

Supplier: TCI America


D2671-5G
TCD2671-005GEA 537.89 USD InStock
TCD2671-005G
2,3-Dibromo-1,1,3,3-tetrafluoropropene ≥95.0%
2,3-Dibromo-1,1,3,3-tetrafluoropropene
CAS Number: 666-40-0
MDL Number: MFCD00236663
Molecular Formula: C3Br2F4
Molecular Weight: 271.83
Purity/Analysis Method: 95.0% (GC)
Form: Clear Liquid
Specific Gravity (20/20): 2.20

Product Categories: Synthetic Organic Chemistry, Fluorinated Building Blocks
Other Product Names / Synonyms: 2,3-Dibromoperfluoropropene, Perfluoro-2,3-dibromopropene, 1,1,3,3-Tetrafluoro-2,3-dibromopropene


Formula: C₃Br₂F₄
Density: 2.20 (20 °C)
MDL Number: MFCD00236663
CAS Number: 666-40-0
EINECS: 000-000-0
Order Now
Specification Test Results
Purity/Analysis Method 95.0% (GC)


 

ORDER

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty