2,4-Dimethyl-6-hydroxypyrimidine ≥98.0% (by HPLC, titration analysis)
Supplier: TCI America
Synonyms:
6-Hydroxy-2,4-dimethylpyrimidine
, 2,6-Dimethylpyrimidin-4-ol
, 2,6-Dimethyl-4-pyrimidinol
D2579-5G
D2579-25G
TCD2579-025GEA
350
USD
TCD2579-025G
TCD2579-005G
2,4-Dimethyl-6-hydroxypyrimidine ≥98.0% (by HPLC, titration analysis)
2,4-Dimethyl-6-hydroxypyrimidine
CAS Number: 6622-92-0
MDL Number: MFCD00006105
Molecular Formula: C6H8N2O
Molecular Weight: 124.14
Purity/Analysis Method:
Form: Crystal
Melting point (°C): 199
Other Product Names / Synonyms: 2,6-Dimethyl-4-pyrimidinol, 6-Hydroxy-2,4-dimethylpyrimidine
Formula:
C₆H₈N₂O MW: 124.14 g/mol Melting Pt: 199 °C |
MDL Number:
MFCD00006105 CAS Number: 6622-92-0 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...