2,4-Dichloro-5-sulfamoylbenzoic acid ≥97.0% (by HPLC, titration analysis)
Supplier: TCI America
D2541-25G
TCD2541-025GEA
35.99
USD
TCD2541-025G
2,4-Dichloro-5-sulfamoylbenzoic acid ≥97.0% (by HPLC, titration analysis)
2,4-Dichloro-5-sulphamoylbenzoic acid
CAS Number: 2736-23-4
MDL Number: MFCD00007931
Molecular Formula: C7H5Cl2NO4S
Molecular Weight: 270.08
Purity/Analysis Method:
Form: Crystal
Other Product Names / Synonyms: 5-(Aminosulfonyl)-2,4-dichlorobenzoic Acid
Formula:
C₇H₅Cl₂NO₄S MW: 270.09 g/mol |
MDL Number:
MFCD00007931 CAS Number: 2736-23-4 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...