(S)-N,N-Dimethyl-1-[(R)-2-(diphenylphosphino)ferrocenyl]ethylamine ≥97.0% (by titrimetric analysis)

Supplier: TCI America

Synonyms: (S)-(R)-PPFA

D2536-100MG
TCD2536-100MGEA 57.84 USD
TCD2536-100MG
(S)-N,N-Dimethyl-1-[(R)-2-(diphenylphosphino)ferrocenyl]ethylamine ≥97.0% (by titrimetric analysis)
(S)-N,N-Dimethyl-1-[(R)-2-(diphenylphosphino)ferrocenyl]ethylamine

CAS Number: 55650-58-3
MDL Number: MFCD00075285
Molecular Formula: C26H28FeNP
Molecular Weight: 441.34
Purity/Analysis Method: 97.0% (T)
Form: Crystal
Melting point (°C): 145


Product Categories: Synthetic Organic Chemistry, Asymmetric Synthesis, Cross-coupling Reaction using Transition Metal Catalysts, Ligands
Other Product Names / Synonyms: (S)-(R)-PPFA

Formula: C₂₆H₂₈FeNP
MW: 441.34 g/mol
Melting Pt: 145 °C
MDL Number: MFCD00075285
CAS Number: 55650-58-3
https://us.vwr.com/stibo/web/std.lang.all/95/06/28979506.jpg

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR