TCIAD2527

D2527-25G
TCD2527-025GEA 69.03 USD
TCD2527-025G
TCIAD2527
CAS Number: 136997-71-2
MDL Number: MFCD00185917
Molecular Formula: C10H14N2O4S
Molecular Weight: 258.29
Form: Crystal
Color: Gray

Other Product Names / Synonyms: 2-[1-(Dimethylcarbamoyl)-4-pyridinio]ethylsulfonate
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR