Methyldiphenylphosphine oxide ≥98.0%

Supplier: TCI America

D2445-10G
TCD2445-010GEA 81 USD
TCD2445-010G
Methyldiphenylphosphine oxide ≥98.0%
Methyldiphenylphosphine oxide

CAS Number: 2129-89-7
MDL Number: MFCD00002081
Molecular Formula: C13H13OP
Molecular Weight: 216.22
Purity/Analysis Method: 98.0% (GC)
Form: Crystal
Boiling point (°C): 180
Melting point (°C): 113

Formula: C₁₃H₁₃OP
MW: 216.22 g/mol
Boiling Pt: 180 °C
Melting Pt: 113 °C
MDL Number: MFCD00002081
CAS Number: 2129-89-7
https://us.vwr.com/stibo/web/std.lang.all/93/69/28979369.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR