Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

trans-2,4-Difluorocinnamic acid ≥98.0% (by GC, titration analysis)

Supplier: TCI America

Synonyms: 2,4-Difluorocinnamic acid

D2343-5G
TCD2343-005GEA 83.23 USD InStock
TCD2343-005G
trans-2,4-Difluorocinnamic acid ≥98.0% (by GC, titration analysis)
trans-2,4-Difluorocinnamic acid
CAS Number: 94977-52-3
MDL Number: MFCD00010317
Molecular Formula: C9H6F2O2
Molecular Weight: 184.14
Purity/Analysis Method: 98.0% (GC,T)
Form: Crystal
Melting point (°C): 217

Product Categories: Synthetic Organic Chemistry, Fluorinated Building Blocks
Other Product Names / Synonyms: 3-(2,4-Difluorophenyl)acrylic Acid


Formula: C₉H₆F₂O₂
MW: 184.14 g/mol
Melting Pt: 217 °C
MDL Number: MFCD00010317
CAS Number: 94977-52-3
EINECS: 000-000-0
Order Now
Specification Test Results
Purity/Analysis Method 98.0% (GC,T)


 

ORDER

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty