Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4,4'-Diacetoxybiphenyl

Supplier: TCI America

D2266-25G
TCD2266-025G
4,4'-Diacetoxybiphenyl
CAS Number: 32604-29-8
MDL Number: MFCD00035973
Molecular Formula: C16H14O4
Molecular Weight: 270.28
Form: Crystal
Color: White
Melting point (°C): 164


Formula: C₁₆H₁₄O₄
MW: 270,28
Melting Pt: 164 °C
MDL Number: MFCD00035973
CAS Number: 32604-29-8
EINECS: 000-000-0
Order Now
Specification Test Results
Melting point (°C)
Color White


 

ORDER

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty