1,3-Diphenylacetone-p-toluenesulfonylhydrazone ≥98.0% (by HPLC, titration analysis)
Supplier: TCI America
D2093-10G
D2093-25G
TCD2093-010GEA
60.9
USD
TCD2093-010G
TCD2093-25G
1,3-Diphenylacetone-p-toluenesulfonylhydrazone ≥98.0% (by HPLC, titration analysis)
1,3-Diphenylacetone p-toluenesulphonylhydrazone
CAS Number: 19816-88-7
MDL Number: MFCD00009645
Molecular Formula: C22H22N2O2S
Molecular Weight: 378.49
Purity/Analysis Method:
Form: Crystal
Melting point (°C): 185
Other Product Names / Synonyms: 1,3-Diphenylacetone p-Tosylhydrazone, 1,3-Diphenyl-2-propanone Tosylhydrazone
Formula:
C₂₂H₂₂N₂O₂S Melting Pt: 185 °C |
MDL Number:
MFCD00009645 CAS Number: 19816-88-7 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...