5,8-Dihydroxy-1,4-naphthoquinone ≥75.0% (by HPLC)

Supplier: TCI America

D2070-1G
TCD2070-001GEA 407.02 USD
TCD2070-001G
5,8-Dihydroxy-1,4-naphthoquinone ≥75.0% (by HPLC)
5,8-Dihydroxy-1,4-naphthoquinone

CAS Number: 475-38-7
MDL Number: MFCD00001685
Molecular Formula: C10H6O4
Molecular Weight: 190.15
Purity/Analysis Method: 75.0% (HPLC)
Form: Crystal
Melting point (°C): 230


Other Product Names / Synonyms: 5,8-Dihydroxy-1,4-naphthalenedione, Naphthazarin

https://us.vwr.com/stibo/web/std.lang.all/88/54/28978854.jpg

Order Now

Specification Test Results

Purity/Analysis Method 75.0% (HPLC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR