N,N-Dibenzylhydroxylamine ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

D2004-25G
TCD2004-025GEA 109.54 USD
TCD2004-025G
N,N-Dibenzylhydroxylamine ≥98.0% (by HPLC, titration analysis)
N,N-Dibenzylhydroxylamine

CAS Number: 621-07-8
MDL Number: MFCD00004772
Molecular Formula: C14H15NO
Molecular Weight: 213.28
Purity/Analysis Method: 98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 124
Flash Point (°C): 100

https://us.vwr.com/stibo/web/std.lang.all/87/95/28978795.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR