1,5,9-Decatriene ≥95.0%

Supplier: TCI America

D1993-25ML
TCD1993-025MLEA 169 USD
TCD1993-025ML
1,5,9-Decatriene ≥95.0%
1,5,9-Decatriene

CAS Number: 13393-64-1
MDL Number: MFCD00055625
Molecular Formula: C10H16
Molecular Weight: 136.24
Purity/Analysis Method: 95.0% (GC)
Form: Clear Liquid
Boiling point (°C): 55
Flash Point (°C): 46
Specific Gravity (20/20): 0.77

Formula: C₁₀H₁₆
Boiling Pt: 55 °C
Density: 0.77 (20 °C)
Flash Pt: 46 °C
MDL Number: MFCD00055625
CAS Number: 13393-64-1
https://us.vwr.com/stibo/web/std.lang.all/87/87/28978787.jpg

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR