(Dimethylamino)acetonitrile ≥96.0% (by GC, titration analysis)

Supplier: TCI America

D1929-25ML
TCD1929-25MLEA 58.3 USD
TCD1929-25ML
(Dimethylamino)acetonitrile ≥96.0% (by GC, titration analysis)
(Dimethylamino)acetonitrile

CAS Number: 926-64-7
MDL Number: MFCD00001890
Molecular Formula: C4H8N2
Molecular Weight: 84.12
Purity/Analysis Method: 96.0% (GC,T)
Form: Clear Liquid
Boiling point (°C): 138
Flash Point (°C): 36
Specific Gravity (20/20): 0.87

https://us.vwr.com/stibo/web/std.lang.all/87/29/28978729.jpg

Order Now

Specification Test Results

Purity/Analysis Method 96.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR