3,5-Dichloro-2-hydroxybenzenesulfonic acid sodium salt ≥99.0% (by titrimetric analysis) for biochemical research
Supplier: TCI America
Synonyms:
Sodium 3,5-Dichloro-2-hydroxybenzenesulphonate
, Sodium 3,5-Dichloro-2-hydroxybenzenesulfonate
D1928-25G
TCD1928-025GEA
114.7
USD
TCD1928-025G
3,5-Dichloro-2-hydroxybenzenesulfonic acid sodium salt ≥99.0% (by titrimetric analysis) for biochemical research
3,5-Dichloro-2-hydroxybenzenesulphonic acid sodium salt
[for Biochemical Research]
CAS Number: 54970-72-8
MDL Number: MFCD00009798
Molecular Formula: C6H4Cl2O4S
Molecular Weight: 265.04
Purity/Analysis Method:
Form: Crystal
Product Categories: Bioscience, Peroxidase Substrates
Other Product Names / Synonyms: 2,4-Dichlorophenol-6-sulfonic Acid Sodium Salt, 3,5-Dichloro-2-hydroxybenzenesulfonic Acid Sodium Salt
Formula:
C₆H₃Cl₂NaO₄S MW: 265.05 g/mol |
MDL Number:
MFCD00009798 CAS Number: 54970-72-8 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...