(+)-Diacetyl-L-tartaric anhydride ≥97.0% (by titrimetric analysis)

Supplier: TCI America

D1911-25G
TCD1911-025GEA 59 USD
TCD1911-025G
(+)-Diacetyl-L-tartaric anhydride ≥97.0% (by titrimetric analysis)
(+)-Diacetyl-L-tartaric anhydride

CAS Number: 6283-74-5
MDL Number: MFCD00037918
Molecular Formula: C8H8O7
Molecular Weight: 216.15
Purity/Analysis Method: 97.0% (T)
Form: Crystal
Melting point (°C): 134
Specific rotation [a]20/D: 95 deg (C=0.5, CHCl3)
Storage Temperature: 0°C

Formula: C₈H₈O₇
Melting Pt: 134 °C
MDL Number: MFCD00037918
CAS Number: 6283-74-5
https://us.vwr.com/stibo/web/std.lang.all/87/09/28978709.jpg

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR