Methyl-3,3-dimethoxypropionate ≥98.0%

Supplier: TCI America

Synonyms: Methyl 3,3-dimethoxypropanoate

D1858-500G D1858-25G
TCD1858-500GEA 538.35 USD
TCD1858-500G TCD1858-025G
Methyl-3,3-dimethoxypropionate ≥98.0%
Methyl 3,3-dimethoxypropionate

CAS Number: 7424-91-1
MDL Number: MFCD00010650
Molecular Formula: C6H12O4
Molecular Weight: 148.16
Purity/Analysis Method: 98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 80
Flash Point (°C): 67
Specific Gravity (20/20): 1.06


Other Product Names / Synonyms: 3,3-Dimethoxypropionic Acid Methyl Ester

https://us.vwr.com/stibo/web/std.lang.all/86/55/28978655.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR