1,3-Dichloro-1,1,3,3-tetraisopropyldisiloxane ≥97.0%

Supplier: TCI America

Synonyms: 1,3-Dichloro-1,1,3,3-tetrakis(isopropyl)disiloxane

D1608-5G D1608-25G
TCD1608-25GEA 353.43 USD
TCD1608-25G TCD1608-005G
1,3-Dichloro-1,1,3,3-tetraisopropyldisiloxane ≥97.0%
1,3-Dichloro-1,1,3,3-tetraisopropyldisiloxane

[Hydroxyl Protecting Agent]
CAS Number: 69304-37-6
MDL Number: MFCD00009655
Molecular Formula: C12H28Cl2OSi2
Molecular Weight: 315.43
Purity/Analysis Method: 97.0% (GC)
Form: Clear Liquid
Boiling point (°C): 90
Flash Point (°C): 76
Specific Gravity (20/20): 1.01

Formula: C₁₂H₂₈Cl₂OSi₂
MW: 315.43
Boiling Pt: 90 °C
Density: 1.01 (20 °C)
Flash Pt: 76 °C
MDL Number: MFCD00009655
CAS Number: 69304-37-6
UN: 1760
ADR: 8,II
https://us.vwr.com/stibo/web/std.lang.all/84/07/28978407.jpg

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR