2,5-Diaminotoluene sulfate ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

Synonyms: 2-Methyl-1,4-pHenylenediammonium sulphate2-Methyl-1,4-pHenylenediammonium sulfate

D1602-500G D1602-25G
TCD1602-500GEA 296 USD
TCD1602-500G TCD1602-25G
2,5-Diaminotoluene sulfate ≥98.0% (by HPLC, titration analysis)
2,5-Diaminotoluene sulphate

CAS Number: 615-50-9
MDL Number: MFCD00013003
Molecular Formula: C7H10N2
Molecular Weight: 220.24
Purity/Analysis Method: 98.0% (HPLC,T)
Form: Crystal


Other Product Names / Synonyms: 2,5-Toluenediamine Sulfate, Tolylene-2,5-diamine Sulfate

Formula: C₇H₁₂N₂O₄S
MDL Number: MFCD00013003
UN: 2811
ADR: 6.1,III
https://us.vwr.com/stibo/web/std.lang.all/84/03/28978403.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR