Chlorodimethylpropylsilane ≥99.0%

Supplier: TCI America

Synonyms: DimethylpropylchlorosilanePropyldimethylchlorosilane

D1590-25ML D1590-5ML
TCD1590-25MLEA 81.09 USD
TCD1590-25ML TCD1590-005ML
Chlorodimethylpropylsilane ≥99.0%
Chlorodimethylpropylsilane

[Dimethylpropylsilylating Agent]
CAS Number: 17477-29-1
MDL Number: MFCD00039301
Molecular Formula: C5H13ClSi
Molecular Weight: 136.69
Purity/Analysis Method: 99.0% (GC)
Form: Clear Liquid
Boiling point (°C): 116
Flash Point (°C): 24
Specific Gravity (20/20): 0.87


Product Categories: Materials Chemistry, Silane Coupling Agents, Analytical Science, Silylation (GC Derivatizing Reagents)
Other Product Names / Synonyms: Dimethylpropylchlorosilane, Propyldimethylchlorosilane

Formula: C₅H₁₃ClSi
MW: 136.69 g/mol
Boiling Pt: 116 °C
Density: 0.87 (20 °C)
Flash Pt: 24 °C
MDL Number: MFCD00039301
CAS Number: 17477-29-1
https://us.vwr.com/stibo/web/std.lang.all/83/91/28978391.jpg

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR