Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Dimetridazole ≥98.0% (by GC, titration analysis)

Supplier: TCI America
Synonyms: 1,2-Dimethyl-5-nitroimidazole

D1564-25G D1564-500G
TCD1564-025G TCD1564-500G
Dimetridazole ≥98.0% (by GC, titration analysis)
CAS Number: 551-92-8
MDL Number: MFCD00047046
Molecular Formula: C5H7N3O2
Molecular Weight: 141.13
Purity/Analysis Method: 98.0% (GC,T)
Form: Crystal
Melting point (°C): 140
Storage Temperature: 0-10°C

Product Categories: Bioscience, Antibiotic Ingredients, Anthelmintic Ingredients
Other Product Names / Synonyms: Dimetridazole


Formula: C₅H₇N₃O₂
MW: 141,13
Melting Pt: 140 °C
MDL Number: MFCD00047046
CAS Number: 551-92-8
EINECS: 209-001-2
Order Now
Specification Test Results
Purity/Analysis Method >98.0% (GC,T)
Melting point (°C) 140
Storage Temperature 0-10°C


 

ORDER

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty