Chlorodimethyloctadecylsilane ≥97.0% (by titrimetric analysis)

Supplier: TCI America

D1560-25ML
TCD1560-025MLEA 132.97 USD
TCD1560-025ML
Chlorodimethyloctadecylsilane ≥97.0% (by titrimetric analysis)
Chlorodimethyloctadecylsilane

CAS Number: 18643-08-8
MDL Number: MFCD00000504
Molecular Formula: C20H43ClSi
Molecular Weight: 347.10
Purity/Analysis Method: 97.0% (T)
Form: Crystal
Flash Point (°C): 201
Freezing point (°C): 29


Product Categories: Materials Chemistry, Silane Coupling Agents
Other Product Names / Synonyms: Chlorodimethyloctadecylsilane, n-Octadecyldimethylchlorosilane, Stearyldimethylchlorosilane

Formula: C₂₀H₄₃ClSi
Flash Pt: 201 °C
MDL Number: MFCD00000504
CAS Number: 18643-08-8
https://us.vwr.com/stibo/web/std.lang.all/83/65/28978365.jpg

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR