N,N'-Diallyl-L-tartardiamide ≥98.0% (by titrimetric analysis)
Supplier: TCI America
D1539-25G
TCD1539-025GEA
84.21
USD
TCD1539-025G
N,N'-Diallyl-L-tartardiamide ≥98.0% (by titrimetric analysis)
N,N'-Diallyl-L-tartardiamide
CAS Number: 58477-85-3
MDL Number: MFCD00008640
Molecular Formula: C10H16N2O4
Molecular Weight: 228.25
Purity/Analysis Method:
Form: Crystal
Melting point (°C): 183
Specific rotation [a]20/D: 108 deg (C=2.4, H2O)
Product Categories: Synthetic Organic Chemistry, Chiral Building Blocks, Materials Chemistry, Monomers
Other Product Names / Synonyms: (+)-N,N'-Diallyl-L-tartramide, DATD
Formula:
C₁₀H₁₆N₂O₄ Melting Pt: 183 °C |
MDL Number:
MFCD00008640 CAS Number: 58477-85-3 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...