Dihydrojasmone ≥98.0%

Supplier: TCI America

Synonyms: 3-Methyl-2-pentylcyclopent-2-en-1-one

D1440-25ML
TCD1440-025MLEA 27 USD
TCD1440-025ML
Dihydrojasmone ≥98.0%
Dihydrojasmone

CAS Number: 1128-08-1
MDL Number: MFCD00036480
Molecular Formula: C11H18O
Molecular Weight: 166.26
Purity/Analysis Method: 98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 84
Flash Point (°C): 96
Specific Gravity (20/20): 0.92


Other Product Names / Synonyms: 2-Amyl-3-methyl-2-cyclopenten-1-one

Formula: C₁₁H₁₈O
MW: 166.26 g/mol
Boiling Pt: 84 °C
Density: 0.92 (20 °C)
Flash Pt: 96 °C
MDL Number: MFCD00036480
CAS Number: 1128-08-1
https://us.vwr.com/stibo/web/std.lang.all/82/74/28978274.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR