N,N-Diethyldodecanamide ≥98.0%

Supplier: TCI America

Synonyms: N-LauroyldiethylamineN-DodecanoyldiethylamineN,N-Diethyllauramide

D1407-25ML D1407-500ML
TCD1407-025MLEA 32.46 USD
TCD1407-025ML TCD1407-500ML
N,N-Diethyldodecanamide ≥98.0%
N,N-Diethyldodecanamide

CAS Number: 3352-87-2
MDL Number: MFCD00008962
Molecular Formula: C16H33NO
Molecular Weight: 255.45
Purity/Analysis Method: 98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 167
Flash Point (°C): 113
Freezing point (°C): 2
Specific Gravity (20/20): 0.87


Other Product Names / Synonyms: N,N-Diethyllauramide, N-Dodecanoyldiethylamine, N-Lauroyldiethylamine

Formula: C₁₆H₃₃NO
MW: 255.45 g/mol
Boiling Pt: 167 °C
Density: 0.87 (20 °C)
Flash Pt: 113 °C
MDL Number: MFCD00008962
CAS Number: 3352-87-2
https://us.vwr.com/stibo/web/std.lang.all/82/51/28978251.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR