N,N'-Diacetylethylenediamine ≥98.0% (by GC, titration analysis)

Supplier: TCI America

Synonyms: N,N'-Ethylenebisacetamide

D1347-25G
TCD1347-025GEA 89.32 USD
TCD1347-025G
N,N'-Diacetylethylenediamine ≥98.0% (by GC, titration analysis)
N,N'-Diacetylethylenediamine

CAS Number: 871-78-3
MDL Number: MFCD00154671
Molecular Formula: C6H12N2O2
Molecular Weight: 144.17
Purity/Analysis Method: 98.0% (GC,T)
Form: Crystal
Melting point (°C): 175


Other Product Names / Synonyms: N,N'-Ethylenebisacetamide

Formula: C₆H₁₂N₂O₂
MW: 144.17 g/mol
Melting Pt: 175 °C
MDL Number: MFCD00154671
CAS Number: 871-78-3
https://us.vwr.com/stibo/web/std.lang.all/82/12/28978212.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR