3-Decyn-1-ol ≥97.0%
Supplier: TCI America
D1275-5ML
TCD1275-005MLEA
80.7
USD
TCD1275-005ML
3-Decyn-1-ol ≥97.0%
3-Decyn-1-ol
CAS Number: 51721-39-2
MDL Number: MFCD00040917
Molecular Formula: C10H18O
Molecular Weight: 154.25
Purity/Analysis Method:
Form: Clear Liquid
Boiling point (°C): 131
Flash Point (°C): 107
Specific Gravity (20/20): 0.87
Other Product Names / Synonyms: Hexyl(2-hydroxyethyl)acetylene
Formula:
C₁₀H₁₈O Boiling Pt: 131 °C Density: 0.87 (20 °C) Flash Pt: 107 °C |
MDL Number:
MFCD00040917 CAS Number: 51721-39-2 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...