Diethyl dibutylmalonate ≥98.0%

Supplier: TCI America

Synonyms: Diethyl 2,2-dibutylmalonate

D1184-25ML
TCD1184-025MLEA 76 USD
TCD1184-025ML
Diethyl dibutylmalonate ≥98.0%
Diethyl dibutylmalonate

CAS Number: 596-75-8
MDL Number: MFCD00026842
Molecular Formula: C15H28O4
Molecular Weight: 272.39
Purity/Analysis Method: 98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 142
Specific Gravity (20/20): 0.95


Other Product Names / Synonyms: Dibutylmalonic Acid Diethyl Ester

https://us.vwr.com/stibo/web/std.lang.all/81/05/28978105.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR