Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Diethyl Diallylmalonate ≥98.0%

Supplier: TCI America

Synonyms: Diallylmalonic Acid Diethyl Ester

D1183-25ML
TCD1183-025MLEA 97.8 USD InStock
TCD1183-025ML
Diethyl Diallylmalonate ≥98.0%
Diethyl diallylmalonate
CAS Number: 3195-24-2
MDL Number: MFCD00009126
Molecular Formula: C13H20O4
Molecular Weight: 240.30
Purity/Analysis Method: 98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 128
Flash Point (°C): 113
Specific Gravity (20/20): 0.99

Product Categories: Materials Chemistry, Monomers
Other Product Names / Synonyms: Diallylmalonic Acid Diethyl Ester


Formula: C₁₃H₂₀O₄
MW: 240.30 g/mol
Boiling Pt: 128 °C
Density: 0.99 (20 °C)
Flash Pt: 113 °C
MDL Number: MFCD00009126
CAS Number: 3195-24-2
Order Now
Specification Test Results
Purity/Analysis Method 98.0% (GC)


 

ORDER

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty