4-(Diethylamino)azobenzene ≥97.0% (by HPLC, titration analysis)

Supplier: TCI America

D1126-100G
TCD1126-100GEA 138.44 USD
TCD1126-100G
4-(Diethylamino)azobenzene ≥97.0% (by HPLC, titration analysis)
4-(Diethylamino)azobenzene

CAS Number: 2481-94-9
MDL Number: MFCD00009043
Molecular Formula: C16H19N3
Molecular Weight: 253.35
Purity/Analysis Method: 97.0% (HPLC,T)
Form: Crystal
Melting point (°C): 97


Other Product Names / Synonyms: Solvent Yellow 56, N,N-Diethyl-4-(phenylazo)aniline

Formula: C₁₆H₁₉N₃
Melting Pt: 97 °C
MDL Number: MFCD00009043
CAS Number: 2481-94-9

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (HPLC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR