Quinoxaline-2,3-dione ≥98.0% (by titrimetric analysis)

Supplier: TCI America

Synonyms: 2,3-DihydroxyquinoxalineQuinoxaline-2,3(1H,4H)-dione

D1010-25G
TCD1010-025GEA 60.9 USD
TCD1010-025G
Quinoxaline-2,3-dione ≥98.0% (by titrimetric analysis)
Quinoxaline-2,3-dione

CAS Number: 15804-19-0
MDL Number: MFCD00006723
Molecular Formula: C8H6N2O2
Molecular Weight: 162.15
Purity/Analysis Method: 98.0% (T)
Form: Crystal


Other Product Names / Synonyms: 2,3-Dihydroxyphenpiazine, 2,3-Quinoxalinediol

Formula: C₈H₆N₂O₂
MW: 162.15 g/mol
MDL Number: MFCD00006723
CAS Number: 15804-19-0
https://us.vwr.com/stibo/web/std.lang.all/61/99/28976199.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR