1,2-Diphenoxyethane ≥99.0%

Supplier: TCI America

D0866-5G
TCD0866-005GEA 46.7 USD
TCD0866-005G
1,2-Diphenoxyethane ≥99.0%
1,2-Diphenoxyethane

CAS Number: 104-66-5
MDL Number: MFCD00003039
Molecular Formula: C14H14O2
Molecular Weight: 214.26
Purity/Analysis Method: 99.0% (GC)
Form: Crystal
Boiling point (°C): 185
Melting point (°C): 97


Product Categories: Materials Chemistry, Pressure & Heat Sensitive Recording Materials
Other Product Names / Synonyms: 1,2-Diphenoxyethane, Diphenyl Cellosolve, Diphenyl Glycol

Formula: C₁₄H₁₄O₂
MW: 214.26 g/mol
Boiling Pt: 185 °C
Melting Pt: 97 °C
MDL Number: MFCD00003039
CAS Number: 104-66-5
https://us.vwr.com/stibo/web/std.lang.all/59/67/28975967.jpg

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR