Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

1,3-Di(o-tolyl)-2-thiourea ≥98.0% (by HPLC, total nitrogen)

Supplier: TCI America
Synonyms: N,N'-Di-o-tolylthiourea

D0802-25G
TCD0802-025G
1,3-Di(o-tolyl)-2-thiourea ≥98.0% (by HPLC, total nitrogen)
CAS Number: 137-97-3
MDL Number: MFCD00025922
Molecular Formula: C15H16N2S
Molecular Weight: 256.37
Purity/Analysis Method: 98.0% (HPLC,N)
Form: Crystal
Melting point (°C): 152


Formula: C₁₅H₁₆N₂S
MW: 256,37
Melting Pt: 152 °C
MDL Number: MFCD00025922
CAS Number: 137-97-3
Order Now
Specification Test Results
Purity/Analysis Method >98.0% (HPLC,N)
Melting point (°C) 152


 

ORDER

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty