Methyl dimethoxyacetate ≥95.0%

Supplier: TCI America

Synonyms: Methyl 2,2-dimethoxyacetate

D0503-25ML
TCD0503-025MLEA 109.62 USD
TCD0503-025ML
Methyl dimethoxyacetate ≥95.0%
Methyl dimethoxyacetate

CAS Number: 89-91-8
MDL Number: MFCD00008484
Molecular Formula: C5H10O4
Molecular Weight: 134.13
Purity/Analysis Method: 95.0% (GC)
Form: Clear Liquid
Boiling point (°C): 67
Flash Point (°C): 67
Specific Gravity (20/20): 1.10


Other Product Names / Synonyms: Dimethoxyacetic Acid Methyl Ester

Formula: C₅H₁₀O₄
MW: 134.13 g/mol
Boiling Pt: 67 °C
Density: 1.10 (20 °C)
Flash Pt: 67 °C
MDL Number: MFCD00008484
CAS Number: 89-91-8
https://us.vwr.com/stibo/web/std.lang.all/50/87/28975087.jpg

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR