4,6-Dichlororesorcinol ≥96.0%
Supplier: TCI America
D0413-25G
TCD0413-025GEA
202.94
USD
TCD0413-025G
4,6-Dichlororesorcinol ≥96.0%
4,6-Dichlororesorcinol
CAS Number: 137-19-9
MDL Number: MFCD00002274
Molecular Formula: C6H4Cl2O2
Molecular Weight: 179.00
Purity/Analysis Method:
Form: Crystal
Boiling point (°C): 125
Melting point (°C): 115
Other Product Names / Synonyms: 4,6-Dichloro-1,3-dihydroxybenzene
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...