1,3-Di-4-piperidylpropane ≥97.0%
Supplier: TCI America
Synonyms:
4,4'-Trimethylenedipiperidine
, 1,3-Bis(4-piperidyl)propane
D0293-25G
D0293-500G
TCD0293-025GEA
44.45
USD
TCD0293-025G
TCD0293-500G
1,3-Di-4-piperidylpropane ≥97.0%
1,3-Di-4-piperidylpropane
CAS Number: 16898-52-5
MDL Number: MFCD00038013
Molecular Formula: C13H26N2
Molecular Weight: 210.37
Purity/Analysis Method:
Form: Crystal
Melting point (°C): 66
Flash Point (°C): 137
Other Product Names / Synonyms: 1,3-Bis(4-piperidyl)propane, 4,4'-Trimethylenedipiperidine
Formula:
C₁₃H₂₆N₂ MW: 210.37 g/mol Melting Pt: 66 °C Flash Pt: 137 °C |
MDL Number:
MFCD00038013 CAS Number: 16898-52-5 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...