meso-1,2-Dibromo-1,2-diphenylethane ≥97.0% (by titrimetric analysis)

Supplier: TCI America

D0183-25G
TCD0183-025GEA 119 USD
TCD0183-025G
meso-1,2-Dibromo-1,2-diphenylethane ≥97.0% (by titrimetric analysis)
meso-1,2-Dibromo-1,2-diphenylethane

CAS Number: 13440-24-9
MDL Number: MFCD00085299
Molecular Formula: C14H12Br2
Molecular Weight: 340.06
Purity/Analysis Method: 97.0% (T)
Form: Crystal


Other Product Names / Synonyms: meso-Stilbene Dibromide

Formula: C₁₄H₁₂Br₂
MDL Number: MFCD00000137
CAS Number: 13440-24-9

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR