Diacetone acrylamide ≥98.0% (by GC, titration analysis) stabilized

Synonyms: N-(1,1-Dimethyl-3-oxobutyl)acrylamide

D0062-25G
TCD0062-025GEA 28.41 USD
TCD0062-025G
Diacetone acrylamide ≥98.0% (by GC, titration analysis) stabilized
Diacetone acrylamide

(stabilized with MEHQ)
CAS Number: 2873-97-4
MDL Number: MFCD00008788
Molecular Formula: C9H15NO2
Molecular Weight: 169.22
Purity/Analysis Method: 98.0% (GC,T)
Form: Crystal
Boiling point (°C): 120
Melting point (°C): 57
Flash Point (°C): 110


Product Categories: Materials Chemistry, Monomers
Other Product Names / Synonyms: N-(1,1-Dimethyl-3-oxobutyl)acrylamide

Formula: C₉H₁₅NO₂
MW: 169.22 g/mol
Boiling Pt: 120 °C
Melting Pt: 57 °C
Flash Pt: 110 °C
MDL Number: MFCD00008788
CAS Number: 2873-97-4
https://us.vwr.com/stibo/web/std.lang.all/43/99/28974399.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR