Didecyl ether ≥95.0%

Supplier: TCI America

D0035-25ML
TCD0035-025MLEA 146.77 USD
TCD0035-025ML
Didecyl ether ≥95.0%
Didecyl ether

CAS Number: 2456-28-2
MDL Number: MFCD00026539
Molecular Formula: C20H42O
Molecular Weight: 298.56
Purity/Analysis Method: 95.0% (GC)
Form: Clear Liquid
Boiling point (°C): 196
Specific Gravity (20/20): 0.82


Other Product Names / Synonyms: Didecyl Ether

Formula: C₂₀H₄₂O
Boiling Pt: 196 °C
Density: 0,82 (20 °C)
MDL Number: MFCD00026539
CAS Number: 2456-28-2

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR