Cinnamyl propionate ≥97.0%

Synonyms: Propionic Acid Cinnamyl Ester

C2375-25G
TCC2375-25GEA 42.84 USD
TCC2375-25G
Cinnamyl propionate ≥97.0%
Cinnamyl propionate

CAS Number: 103-56-0
MDL Number: MFCD00048610
Molecular Formula: C12H14O2
Molecular Weight: 190.24
Purity/Analysis Method: 97.0% (GC)
Form: Clear Liquid
Boiling point (°C): 289
Flash Point (°C): 100
Specific Gravity (20/20): 1.04


Other Product Names / Synonyms: Propionic Acid Cinnamyl Ester

Formula: C₁₂H₁₄O₂
MW: 190.24 g/mol
Boiling Pt: 289 °C
Density: 1.04 (20 °C)
Flash Pt: 100 °C
MDL Number: MFCD00048610
CAS Number: 103-56-0
https://us.vwr.com/stibo/web/std.lang.all/93/64/28939364.jpg

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR