α-Bromo-5-chloro-2-fluorotoluene ≥97.0%
Supplier: TCI America
Synonyms:
5-Chloro-2-fluorobenzyl bromide
, 2-(Bromomethyl)-4-chloro-1-fluorobenzene
C2360-5G
TCC2360-5GEA
82
USD
TCC2360-5G
α-Bromo-5-chloro-2-fluorotoluene ≥97.0%
α-Bromo-5-chloro-2-fluorotoluene
CAS Number: 71916-91-1
MDL Number: MFCD01631437
Molecular Formula: C7H5BrClF
Molecular Weight: 223.47
Purity/Analysis Method:
Form: Clear Liquid
Boiling point (°C): 45
Specific Gravity (20/20): 1.65
Product Categories: Synthetic Organic Chemistry, Fluorinated Building Blocks
Other Product Names / Synonyms: 1-(Bromomethyl)-5-chloro-2-fluorobenzene
Formula:
C₇H₅BrClF MW: 223.47 g/mol Boiling Pt: 45 °C Density: 1.65 (20 °C) |
MDL Number:
MFCD01631437 CAS Number: 71916-91-1 UN: 3265 ADR: 8,II |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...