3-(Chloromethyl)benzoyl chloride ≥93.0%

Supplier: TCI America

C2314-25G C2314-5G
TCC2314-25GEA 151 USD
TCC2314-25G TCC2314-5G
3-(Chloromethyl)benzoyl chloride ≥93.0%
3-(Chloromethyl)benzoyl chloride

CAS Number: 63024-77-1
MDL Number: MFCD00000682
Molecular Formula: C8H6Cl2O
Molecular Weight: 189.04
Purity/Analysis Method: 93.0% (GC)
Form: Clear Liquid
Boiling point (°C): 150
Melting point (°C): 4
Flash Point (°C): 113
Specific Gravity (20/20): 1.33

Formula: C₈H₆Cl₂O
MW: 189.04 g/mol
Boiling Pt: 150 °C
Melting Pt: 4 °C
Density: 1.33 (20 °C)
Flash Pt: 113 °C
MDL Number: MFCD00000682
CAS Number: 63024-77-1
UN: 3265
ADR: 8,II
https://us.vwr.com/stibo/web/std.lang.all/93/17/28939317.jpg

Order Now

Specification Test Results

Purity/Analysis Method 93.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR