1-Chloronaphthalene ≥97.0%

Supplier: TCI America

Synonyms: Triisodecyl phosphiteα-Chloronaphthalene

C2310-5G
TCC2310-5GEA 97.14 USD
TCC2310-5G
1-Chloronaphthalene ≥97.0%
1-Chloronaphthalene

CAS Number: 90-13-1
MDL Number: MFCD00003874
Molecular Formula: C10H7Cl
Molecular Weight: 162.62
Purity/Analysis Method: 97.0% (GC)
Form: Clear Liquid
Boiling point (°C): 263
Melting point (°C): -20
Flash Point (°C): 121
Specific Gravity (20/20): 1.20

Formula: C₁₀H₇Cl
MW: 162.62 g/mol
Boiling Pt: 263 °C
Melting Pt: –20 °C
Density: 1.20 (20 °C)
Flash Pt: 121 °C
MDL Number: MFCD00003874
CAS Number: 90-13-1
UN: 2810
ADR: 6.1,III
https://us.vwr.com/stibo/web/std.lang.all/93/14/28939314.jpg

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR