α-Bromo-2-chloro-6-fluorotoluene ≥98.0%

Supplier: TCI America

Synonyms: 2-(Bromomethyl)-1-chloro-3-fluorobenzene2-Chloro-6-fluorobenzyl bromide

C2308-25G C2308-5G
TCC2308-25GEA 635 USD
TCC2308-25G TCC2308-5G
α-Bromo-2-chloro-6-fluorotoluene ≥98.0%
α-Bromo-2-chloro-6-fluorotoluene

CAS Number: 68220-26-8
MDL Number: MFCD00040126
Molecular Formula: C7H5BrClF
Molecular Weight: 223.47
Purity/Analysis Method: 98.0% (GC)
Form: Clear Liquid
Flash Point (°C): 110
Specific Gravity (20/20): 1.66


Product Categories: Synthetic Organic Chemistry, Fluorinated Building Blocks
Other Product Names / Synonyms: 2-(Bromomethyl)-1-chloro-3-fluorobenzene

Formula: C₇H₅BrClF
MW: 223.47 g/mol
Density: 1.66 (20 °C)
Flash Pt: 110 °C
MDL Number: MFCD00040126
CAS Number: 68220-26-8
UN: 3265
ADR: 8,II
https://us.vwr.com/stibo/web/std.lang.all/93/11/28939311.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR