3-Chlorocoumarin ≥98.0%

Supplier: TCI America

C2297-1G
TCC2297-1GEA 45.66 USD
TCC2297-1G
3-Chlorocoumarin ≥98.0%
3-Chlorocoumarin

CAS Number: 92-45-5
MDL Number: MFCD00024073
Molecular Formula: C9H5ClO2
Molecular Weight: 180.59
Purity/Analysis Method: 98.0% (GC)
Form: Crystal
Melting point (°C): 123

Formula: C₉H₅ClO₂
Melting Pt: 123 °C
MDL Number: MFCD00024073
CAS Number: 92-45-5
https://us.vwr.com/stibo/web/std.lang.all/93/02/28939302.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR