3-Chlorobenzo[b]thiophene-2-carbonyl chlorid ≥97.0% (by GC, titration analysis)
Supplier: TCI America
Synonyms:
3-Chlorobenzo[b]thiophene-2-carbonyl chloride
C2286-1G
TCC2286-1GEA
62
USD
TCC2286-1G
3-Chlorobenzo[b]thiophene-2-carbonyl chlorid ≥97.0% (by GC, titration analysis)
3-Chlorobenzo[b]thiophene-2-carbonyl chlorid
CAS Number: 21815-91-8
MDL Number: MFCD00053069
Molecular Formula: C9H4Cl2OS
Molecular Weight: 231.09
Purity/Analysis Method:
Form: Crystal
Melting point (°C): 115
Formula:
C₉H₄Cl₂OS MW: 231.10 g/mol Melting Pt: 115 °C |
MDL Number:
MFCD00053069 CAS Number: 21815-91-8 UN: 3261 ADR: 8,II |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...