N-Carbobenzoxy-L-threonine benzyl ester ≥98.0% (by HPLC, total nitrogen)

Supplier: TCI America

Synonyms: Z-Thr-OBzlN-Cbz-L-threonine Benzyl Ester

C2285-5G
TCC2285-5GEA 111 USD
TCC2285-5G
N-Carbobenzoxy-L-threonine benzyl ester ≥98.0% (by HPLC, total nitrogen)
N-Carbobenzoxy-L-threonine benzyl ester

CAS Number: 16597-50-5
MDL Number: MFCD00077041
Molecular Formula: C19H21NO5
Molecular Weight: 343.38
Purity/Analysis Method: 98.0% (HPLC,N)
Form: Crystal
Melting point (°C): 82
Specific rotation [a]20/D: -11 deg (C=8, CHCl3)


Product Categories: Bioscience, Amino Acids (N-Protected), Amino Acids (C-Protected)
Other Product Names / Synonyms: N-Cbz-L-threonine Benzyl Ester, Z-Thr-OBzl

Formula: C₁₉H₂₁NO₅
Melting Pt: 82 °C
MDL Number: MFCD00077041
CAS Number: 16597-50-5

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,N)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR