5-Chloro-2-benzoxazolinone ≥98.0% (by GC, titration analysis)
Supplier: TCI America
Synonyms:
5-Chloro-2(3H)-benzoxazolone
, Chloroxazone
, 5-Chlorobenzo[d]oxazol-2(3H)-one
C2237-25G
TCC2237-25GEA
30.45
USD
TCC2237-25G
5-Chloro-2-benzoxazolinone ≥98.0% (by GC, titration analysis)
5-Chloro-2-benzoxazolinone
CAS Number: 95-25-0
MDL Number: MFCD00005717
Molecular Formula: C7H4ClNO2
Molecular Weight: 169.56
Purity/Analysis Method:
Form: Crystal
Melting point (°C): 191
Other Product Names / Synonyms: 5-Chloro-2-benzoxazolone, 5-Chloro-2-benzoxazolinone, 5-Chloro-2-hydroxybenzoxazole
Formula:
C₇H₄ClNO₂ MW: 169.57 g/mol Melting Pt: 191 °C |
MDL Number:
MFCD00005717 CAS Number: 95-25-0 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...