6-Chloro-2-fluoropurine ≥97.0% (by HPLC, titration analysis)
Supplier: TCI America
Synonyms:
6-Chloro-2-fluoro-9H-purine
C2221-1G
TCC2221-1GEA
114
USD
TCC2221-1G
6-Chloro-2-fluoropurine ≥97.0% (by HPLC, titration analysis)
6-Chloro-2-fluoropurine
CAS Number: 1651-29-2
MDL Number: MFCD08461765
Molecular Formula: C5H2ClFN4
Molecular Weight: 172.55
Purity/Analysis Method:
Form: Crystal
Melting point (°C): 162
Formula:
C₅H₂ClFN₄ MW: 172.55 g/mol Melting Pt: 162 °C |
MDL Number:
MFCD02183557 CAS Number: 1651-29-2 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...