Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

3-Chloro-4-fluoroanisole ≥97.0%

Supplier: TCI America


C2217-5G
TCC2217-5GEA 83.23 USD InStock
TCC2217-5G
3-Chloro-4-fluoroanisole ≥97.0%
3-Chloro-4-fluoroanisole
CAS Number: 202925-07-3
MDL Number: MFCD00070778
Molecular Formula: C7H6ClFO
Molecular Weight: 160.57
Purity/Analysis Method: 97.0% (GC)
Form: Clear Liquid
Boiling point (°C): 189
Flash Point (°C): 79
Specific Gravity (20/20): 1.29


Formula: C₇H₆ClFO
MW: 160,58 g/mol
Boiling Pt: 189 °C
Density: 1,29 (20 °C)
Flash Pt: 79 °C
MDL Number: MFCD00070778
CAS Number: 202925-07-3
EINECS: 000-000-0
Order Now
Specification Test Results
Purity/Analysis Method 97.0% (GC)


 

ORDER

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty